Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031000.3 | NP_112262.1 | 4 | 387 | ||
GenBank mRNA | BC059133.1 | - | 4 | 666 | 6918695 | |
D10854.1 | - | 4 | 387 | |||
FQ212523.1 | - | 4 | 629 | |||
FQ212586.1 | - | 4 | 627 | |||
FQ212919.1 | - | 3 | 337 | |||
FQ213220.1 | - | 3 | 334 | |||
FQ213467.1 | - | 4 | 662 | |||
FQ213483.1 | - | 4 | 658 | |||
FQ213491.1 | - | 4 | 660 | |||
FQ213579.1 | - | 4 | 629 | |||
FQ213776.1 | - | 4 | 629 | |||
FQ214173.1 | - | 4 | 629 | |||
FQ214441.1 | - | 4 | 406 | |||
FQ214642.1 | - | 644 | ||||
FQ214700.1 | - | 4 | 602 | |||
FQ215620.1 | - | 4 | 454 | |||
FQ215772.1 | - | 4 | 629 | |||
FQ218383.1 | - | 4 | 629 | |||
FQ218721.1 | - | 4 | 629 | |||
FQ218729.1 | - | 4 | 629 | |||
FQ220055.1 | - | 2 | 230 | |||
FQ226221.1 | - | 4 | 662 | |||
FQ228132.1 | - | 4 | 662 | |||
FQ228842.1 | - | 4 | 658 | |||
FQ229180.1 | - | 3 | 301 | |||
FQ233156.1 | - | 4 | 662 |