Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035238.2 | NP_001030315.1 | 6 | 513 | ||
NM_031021.3 | NP_112283.1 | 6 | 564 | |||
XM_006256081.2 | XP_006256143.1 | 6 | 600 | |||
GenBank mRNA | BC078807.1 | - | 6 | 513 | 7133064 | |
EV778558.1 | - | 6 | 568 | |||
FQ214437.1 | - | 6 | 580 | |||
FQ224491.1 | - | 6 | 584 | |||
FQ230013.1 | - | 6 | 552 | |||
HE864426.1 | - | 5 | 448 | |||
HE864432.1 | - | 448 | ||||
HE864433.1 | - | 448 | ||||
HE864434.1 | - | 448 |