Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031763.3 | NP_113951.1 | 528 | |||
GenBank mRNA | AF016049.1 | - | 343 | |||
BC072510.1 | - | 528 | 7104733 | |||
FQ224997.1 | - | 252 |