Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC2 SIRNA Assays ›
          Gene Symbol:
          Hdac2
          Gene Name:
          histone deacetylase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 43084870 - 43108198 on Build Rnor_6.0
          UniGene ID:
          Rn.1797
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS331532
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hdac2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053447.1 NP_445899.1 10 1225
          GenBank mRNA AF321130.1 - 2 83
          BC161939.1 - 10 1200

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001527, U31814, AB209190, AK097376, AK092156, and others.
          This siRNA also targets the following transcripts in Mouse: NM_008229, AK135572, BC138517, U31758, AB221663.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to amphetamine
          cardiac muscle hypertrophy
          maintenance of chromatin silencing
          transcription, DNA-templated
          protein deacetylation
          positive regulation of cell proliferation
          epidermal cell differentiation
          positive regulation of epithelial to mesenchymal transition
          positive regulation of receptor biosynthetic process
          negative regulation of neuron projection development
          dendrite development
          histone deacetylation
          hippocampus development
          neuron differentiation
          response to caffeine
          response to lipopolysaccharide
          positive regulation of interleukin-1 production
          positive regulation of tumor necrosis factor production
          circadian regulation of gene expression
          cellular response to heat
          response to nicotine
          response to cocaine
          odontogenesis of dentin-containing tooth
          response to drug
          positive regulation of tyrosine phosphorylation of Stat3 protein
          embryonic digit morphogenesis
          ATP-dependent chromatin remodeling
          negative regulation of apoptotic process
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          behavioral response to ethanol
          positive regulation of oligodendrocyte differentiation
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of protein kinase B signaling
          cardiac muscle cell development
          response to hyperoxia
          negative regulation of cardiac muscle cell proliferation
          regulation of sarcomere organization
          hair follicle placode formation
          negative regulation of dendritic spine development
          eyelid development in camera-type eye
          fungiform papilla formation
          histone H3-K9 modification
          cellular response to hydrogen peroxide
          histone H3 deacetylation
          histone H4 deacetylation
          cellular response to retinoic acid
          cellular response to transforming growth factor beta stimulus
          negative regulation of canonical Wnt signaling pathway
          regulation of protein deacetylation
          cellular response to dopamine
          negative regulation of peptidyl-lysine acetylation
          negative regulation of intrinsic apoptotic signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          core promoter binding
          RNA polymerase II repressing transcription factor binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          deacetylase activity
          enzyme binding
          heat shock protein binding
          chromatin DNA binding
          nucleosomal DNA binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          Krueppel-associated box domain binding
          sequence-specific DNA binding
          poly(A) RNA binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          NAD-dependent histone deacetylase activity (H3-K18 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0