Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017027.1 | NP_058723.1 | 6 | 763 | ||
GenBank mRNA | BC078859.1 | - | 763 | |||
K03242.1 | - | 6 | 763 |