Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145086.1 | NP_659554.1 | 2 | 469 | ||
XM_006236616.1 | XP_006236678.1 | 2 | 517 | |||
XM_006236617.1 | XP_006236679.1 | 2 | 517 | |||
XM_006236618.1 | XP_006236680.1 | 2 | 517 | |||
GenBank mRNA | AF388527.1 | - | 2 | 475 | ||
BC079337.1 | - | 2 | 469 | 7106027 | ||
FQ213573.1 | - | 2 | 512 | |||
FQ214321.1 | - | 2 | 468 | |||
FQ223736.1 | - | 2 | 490 | |||
FQ225988.1 | - | 2 | 184 | |||
FQ226374.1 | - | 2 | 490 | |||
FQ226420.1 | - | 2 | 184 | |||
FQ227636.1 | - | 2 | 490 | |||
FQ228026.1 | - | 2 | 480 | |||
FQ230135.1 | - | 2 | 496 | |||
FQ230382.1 | - | 2 | 183 | |||
FQ231329.1 | - | 2 | 184 | |||
FQ231840.1 | - | 2 | 184 | |||
FQ232288.1 | - | 2 | 490 | |||
FQ232894.1 | - | 2 | 491 | |||
FQ233179.1 | - | 2 | 490 | |||
FQ233457.1 | - | 2 | 496 | |||
FQ233487.1 | - | 2 | 184 | |||
FQ233911.1 | - | 2 | 490 | |||
FQ234284.1 | - | 2 | 464 | |||
FQ234534.1 | - | 2 | 491 | |||
FQ235054.1 | - | 2 | 184 | |||
U21718.1 | - | 1 | 67 |