Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS332496
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
8
843
NM_001034069.1
NP_001029241.1
8
843
NM_001034070.1
NP_001029242.1
8
843
NM_001034071.1
NP_001029243.1
8
843
NM_001034072.1
NP_001029244.1
8
843
NM_001034073.1
NP_001029245.1
7
780
NM_001034074.1
NP_001029246.1
7
780
NM_001034075.1
NP_001029247.1
7
780
NM_001301336.1
NP_001288265.1
8
843
NM_001301342.1
NP_001288271.1
8
843
NM_001301736.1
NP_001288665.1
7
780
NM_019131.2
NP_062004.1
7
780
XM_006243386.2
XP_006243448.1
8
835
XM_006243388.2
XP_006243450.1
8
835
XM_006243389.2
XP_006243451.1
8
835
XM_006243390.2
XP_006243452.1
8
835
XM_006243393.1
XP_006243455.1
7
778
XM_006243394.1
7
796
XM_006243395.2
XP_006243457.1
7
787
XM_008766207.1
XP_008764429.1
8
835
XM_008766208.1
XP_008764430.1
7
778
GenBank mRNA
AA899493.1
-
3
405
AF372215.1
-
7
629
AF372216.1
-
8
742
AF372217.1
-
8
748
BC078780.1
-
7
747
7106337
CF108680.1
-
8
791
FQ214619.1
-
8
818
FQ214803.1
-
8
814
FQ214809.1
-
8
812
FQ214835.1
-
8
812
FQ214864.1
-
8
816
FQ214868.1
-
8
812
FQ214869.1
-
8
818
FQ214872.1
-
8
811
FQ214877.1
-
8
812
FQ214894.1
-
8
818
FQ214911.1
-
8
814
FQ214944.1
-
8
818
FQ214946.1
-
8
818
FQ215107.1
-
8
812
FQ215186.1
-
8
818
FQ215188.1
-
8
812
FQ215206.1
-
8
847
FQ215226.1
-
8
818
FQ215459.1
-
8
814
FQ215512.1
-
8
818
FQ215522.1
-
8
889
FQ215524.1
-
8
814
FQ215538.1
-
8
818
FQ215551.1
-
8
813
FQ215698.1
-
8
812
FQ215703.1
-
8
812
FQ215724.1
-
8
899
FQ215746.1
-
8
814
FQ215761.1
-
8
812
FQ215768.1
-
8
812
FQ215803.1
-
8
818
FQ215808.1
-
8
814
FQ215815.1
-
8
812
FQ215856.1
-
8
812
FQ215919.1
-
8
820
FQ215944.1
-
8
819
FQ215969.1
-
8
812
FQ216014.1
-
8
896
FQ216018.1
-
8
818
FQ216052.1
-
8
812
FQ216082.1
-
8
897
FQ216105.1
-
8
813
FQ216118.1
-
8
896
FQ216145.1
-
8
818
FQ216160.1
-
8
811
FQ216319.1
-
8
814
FQ216368.1
-
8
814
FQ216400.1
-
8
814
FQ216494.1
-
8
812
FQ216552.1
-
8
818
FQ216557.1
-
8
818
FQ216569.1
-
907
FQ216637.1
-
8
814
FQ216683.1
-
8
814
FQ216697.1
-
8
814
FQ216747.1
-
8
814
FQ217468.1
-
7
563
FQ217503.1
-
8
818
FQ217616.1
-
6
380
FQ217637.1
-
8
818
FQ217808.1
-
8
814
FQ223706.1
-
8
814
FQ223993.1
-
8
816
FQ224207.1
-
8
812
FQ224838.1
-
8
818
FQ224943.1
-
8
816
M34134.1
-
7
765
M34135.1
-
8
777
M34136.1
-
6
495
M60666.1
-
8
790
M60667.1
-
8
790
M60668.1
-
7
727
M60669.1
-
7
727
X02411.1
-
8
741
X02412.1
-
6
497
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001164255, NM_001164254, NM_001164256, NM_001164250, NM_024427, and others.
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0