Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS332497
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
3, 4
486
NM_001034069.1
NP_001029241.1
3, 4
486
NM_001034070.1
NP_001029242.1
3, 4
486
NM_001034071.1
NP_001029243.1
3, 4
486
NM_001034072.1
NP_001029244.1
3, 4
486
NM_001034073.1
NP_001029245.1
2, 3
423
NM_001034074.1
NP_001029246.1
2, 3
423
NM_001034075.1
NP_001029247.1
2, 3
423
NM_001301336.1
NP_001288265.1
3, 4
486
NM_001301342.1
NP_001288271.1
3, 4
486
NM_001301736.1
NP_001288665.1
2, 3
423
NM_019131.2
NP_062004.1
2, 3
423
XM_006243386.2
XP_006243448.1
3, 4
478
XM_006243388.2
XP_006243450.1
3, 4
478
XM_006243389.2
XP_006243451.1
3, 4
478
XM_006243390.2
XP_006243452.1
3, 4
478
XM_006243393.1
XP_006243455.1
2, 3
421
XM_006243394.1
2, 3
439
XM_006243395.2
XP_006243457.1
2, 3
430
XM_008766207.1
XP_008764429.1
3, 4
478
XM_008766208.1
XP_008764430.1
2, 3
421
GenBank mRNA
AF372215.1
-
2, 3
272
AF372216.1
-
3, 4
385
AF372217.1
-
3, 4
391
BC078780.1
-
2, 3
390
7106337
BU670983.1
-
3, 4
486
CB585296.1
-
2, 3
426
CF108680.1
-
3, 4
434
CK483405.1
-
3, 4
440
CO562261.1
-
453
FQ074026.1
-
3, 4
544
FQ214619.1
-
3, 4
461
FQ214803.1
-
3, 4
457
FQ214809.1
-
3, 4
455
FQ214835.1
-
3, 4
455
FQ214864.1
-
3, 4
459
FQ214868.1
-
3, 4
455
FQ214869.1
-
3, 4
461
FQ214872.1
-
3, 4
454
FQ214877.1
-
3, 4
455
FQ214894.1
-
3, 4
461
FQ214902.1
-
3, 4
455
FQ214911.1
-
3, 4
457
FQ214944.1
-
3, 4
461
FQ214946.1
-
3, 4
461
FQ214991.1
-
3, 4
455
FQ215186.1
-
3, 4
461
FQ215188.1
-
3, 4
455
FQ215206.1
-
3, 4
490
FQ215226.1
-
3, 4
461
FQ215459.1
-
3, 4
457
FQ215512.1
-
3, 4
461
FQ215522.1
-
3, 4
532
FQ215524.1
-
3, 4
457
FQ215538.1
-
3, 4
461
FQ215551.1
-
3, 4
455
FQ215698.1
-
3, 4
455
FQ215703.1
-
3, 4
455
FQ215724.1
-
3, 4
542
FQ215746.1
-
3, 4
457
FQ215761.1
-
3, 4
455
FQ215768.1
-
3, 4
455
FQ215803.1
-
3, 4
461
FQ215808.1
-
3, 4
457
FQ215815.1
-
3, 4
455
FQ215856.1
-
3, 4
455
FQ215919.1
-
3, 4
463
FQ215944.1
-
3, 4
462
FQ215969.1
-
3, 4
455
FQ215995.1
-
3, 4
457
FQ216014.1
-
3, 4
539
FQ216018.1
-
3, 4
461
FQ216052.1
-
3, 4
455
FQ216105.1
-
3, 4
455
FQ216118.1
-
3, 4
539
FQ216145.1
-
3, 4
461
FQ216160.1
-
3, 4
454
FQ216319.1
-
3, 4
457
FQ216340.1
-
3, 4
461
FQ216368.1
-
3, 4
457
FQ216400.1
-
3, 4
457
FQ216494.1
-
3, 4
455
FQ216552.1
-
3, 4
461
FQ216557.1
-
3, 4
461
FQ216569.1
-
549
FQ216637.1
-
3, 4
457
FQ216683.1
-
3, 4
457
FQ216697.1
-
3, 4
457
FQ216747.1
-
3, 4
457
FQ217468.1
-
2, 3
206
FQ217503.1
-
3, 4
461
FQ217616.1
-
1, 2
23
FQ217637.1
-
3, 4
461
FQ217808.1
-
3, 4
457
FQ223706.1
-
3, 4
457
FQ223993.1
-
3, 4
459
FQ224036.1
-
3, 4
372
FQ224207.1
-
3, 4
455
FQ224838.1
-
3, 4
461
FQ224943.1
-
3, 4
459
FQ224961.1
-
3, 4
457
M34134.1
-
2, 3
408
M34135.1
-
3, 4
420
M34136.1
-
1, 2
138
M60666.1
-
3, 4
433
M60667.1
-
3, 4
433
M60668.1
-
2, 3
370
M60669.1
-
2, 3
370
X02411.1
-
3, 4
384
X02412.1
-
1, 2
140
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001164255, NM_001164254, NM_001164256, NM_001164250, NM_024427, and others.
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0