Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019139.1 | NP_062012.1 | 3 | 441 | ||
XM_006232010.2 | XP_006232072.1 | 3 | 882 | |||
XM_006232011.2 | XP_006232073.1 | 3 | 540 | |||
XM_008760756.1 | XP_008758978.1 | 2 | 1719 | |||
XM_008760757.1 | XP_008758979.1 | 2 | 1641 | |||
GenBank mRNA | AF497634.1 | - | 2 | 369 | ||
EU068467.1 | - | 2 | 329 | |||
EU068468.1 | - | 2 | 329 | |||
EU068469.1 | - | 2 | 407 | |||
EU068470.1 | - | 2 | 392 | |||
EU068471.1 | - | 2 | 392 | |||
EU068472.1 | - | 2 | 392 | |||
L15305.1 | - | 3 | 441 | |||
S75583.1 | - | 2 | 392 |