Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019139.1 | NP_062012.1 | 3 | 412 | ||
XM_006232010.2 | XP_006232072.1 | 3 | 853 | |||
XM_006232011.2 | XP_006232073.1 | 3 | 511 | |||
XM_008760756.1 | XP_008758978.1 | 2 | 1690 | |||
XM_008760757.1 | XP_008758979.1 | 2 | 1612 | |||
GenBank mRNA | AF205713.1 | - | 3 | 416 | ||
AF205714.1 | - | 3 | 338 | |||
AF205715.1 | - | 2 | 239 | |||
AF497634.1 | - | 2 | 340 | |||
EU068467.1 | - | 2 | 300 | |||
EU068468.1 | - | 2 | 300 | |||
EU068469.1 | - | 2 | 378 | |||
EU068470.1 | - | 2 | 363 | |||
EU068471.1 | - | 2 | 363 | |||
EU068472.1 | - | 2 | 363 | |||
L15305.1 | - | 3 | 412 | |||
S75583.1 | - | 2 | 363 |