Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017221.1 | NP_058917.1 | 2 | 629 | ||
GenBank mRNA | AF162915.1 | - | 2 | 63 | ||
L27340.1 | - | 2 | 629 |