Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC4A10 SIRNA Assays ›
          Gene Symbol:
          Slc4a10
          Gene Name:
          solute carrier family 4, sodium bicarbonate transporter, member 10
          Gene Aliases:
          NCBE
          Chromosome Location:
          Chr.3: 47995795 - 48287897 on Build Rnor_6.0
          UniGene ID:
          Rn.15193
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS334607
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc4a10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178092.2 NP_835193.1 20 2626
          XM_006234275.2 XP_006234337.1 20 3177
          XM_006234276.2 XP_006234338.1 20 3174
          XM_006234277.2 XP_006234339.1 20 2957
          XM_006234279.2 XP_006234341.1 20 3172
          XM_006234280.2 XP_006234342.1 19 3084
          XM_006234282.2 XP_006234344.1 19 2867
          XM_006234283.2 XP_006234345.1 19 2864
          XM_006234284.2 XP_006234346.1 19 3083
          XM_006234285.2 XP_006234347.1 19 2866
          GenBank mRNA AF439855.1 - 19 2536
          AF439856.1 - 20 2626
          AY579373.1 - 20 2629
          AY579374.1 - 19 2539
          AY579375.1 - 19 2539
          AY579376.1 - 19 2286
          AY579377.1 - 20 2677
          JX073715.1 - 19 2583
          JX073716.2 - 20 2670
          JX073717.1 - 19 2583
          JX073718.2 - 20 2670
          KF305251.1 - 20 2595
          KF736947.1 - 20 2561
          KF736948.1 - 19 2471
          KF736949.1 - 20 2594
          KF736950.1 - 19 2504
          KJ452197.1 - 21 2610

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          anion transport
          regulation of pH
          nervous system development
          response to light stimulus
          post-embryonic development
          bicarbonate transport
          pyramidal neuron development
          regulation of cellular pH
          ion transmembrane transport
          multicellular organism growth
          locomotory exploration behavior
          sodium ion transmembrane transport
          brain morphogenesis
          regulation of intracellular pH
          anion transmembrane transport
          hydrogen ion transmembrane transport
          inorganic anion exchanger activity
          protein binding
          sodium:bicarbonate symporter activity
          anion:anion antiporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix