Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013165.2 | NP_001013183.2 | 3 | 274 | ||
XM_006237355.2 | XP_006237417.1 | 3 | 316 | |||
XM_006237356.2 | XP_006237418.1 | 3 | 329 | |||
XM_006237358.2 | XP_006237420.1 | 3 | 238 | |||
GenBank mRNA | BC079028.1 | - | 2 | 338 | 7114040 | |
CV118996.1 | - | 3 | 310 | |||
FQ212505.1 | - | 3 | 244 | |||
FQ213657.1 | - | 3 | 244 |
Gene Symbol: LOC100911585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750656.3 | XP_003750704.1 | 3 | 316 | ||
XM_006244627.2 | XP_006244689.1 | 3 | 329 | |||
XM_006244628.2 | XP_006244690.1 | 3 | 238 |