Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT10B SIRNA Assays ›
Gene Symbol:
Wnt10b
Gene Name:
wingless-type MMTV integration site family, member 10B
Gene Aliases:
-
Chromosome Location:
Chr.7: 140448465 - 140454268 on Build Rnor_6.0
UniGene ID:
Rn.218544
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS337280
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Wnt10b
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001108111.1
NP_001101581.1
4
930
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
chondrocyte differentiation
regulation of transcription from RNA polymerase II promoter
lipid metabolic process
cell cycle arrest
smoothened signaling pathway
positive regulation of cell proliferation
positive regulation of G2/M transition of mitotic cell cycle
myoblast differentiation involved in skeletal muscle regeneration
Wnt signaling pathway
neuron differentiation
positive regulation of bone mineralization
positive regulation of epithelial cell differentiation
regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of apoptotic process
skeletal muscle tissue regeneration
cell fate commitment
regulation of fat cell differentiation
negative regulation of fat cell differentiation
positive regulation of osteoblast differentiation
positive regulation of ossification
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
positive regulation of transcription from RNA polymerase II promoter
regulation of skeletal muscle tissue development
skeletal muscle fiber development
negative regulation of epithelial cell proliferation
protein stabilization
sensory perception of taste
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of protein metabolic process
positive regulation of anagen
canonical Wnt signaling pathway
bone trabecula formation
fungiform papilla development
cellular response to organic substance
cellular response to cAMP
cellular response to parathyroid hormone stimulus
cellular response to organic cyclic compound
hematopoietic stem cell proliferation
cellular response to hydrostatic pressure
positive regulation of canonical Wnt signaling pathway
molecular_function
frizzled binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix