Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ISL1 SIRNA Assays ›
          Gene Symbol:
          Isl1
          Gene Name:
          ISL LIM homeobox 1
          Gene Aliases:
          Isl-1, isl-1=homeobox
          Chromosome Location:
          Chr.2: 48488911 - 48501217 on Build Rnor_6.0
          UniGene ID:
          Rn.36202
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS340012
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Isl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017339.3 NP_059035.3 6 961
          GenBank mRNA AY557632.1 - 6 887
          S69329.1 - 6 961

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural crest cell migration
          heart morphogenesis
          secondary heart field specification
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          endocardial cushion morphogenesis
          cardiac right ventricle morphogenesis
          regulation of secondary heart field cardioblast proliferation
          regulation of transcription, DNA-templated
          heart development
          positive regulation of cell proliferation
          regulation of gene expression
          positive regulation of vascular endothelial growth factor production
          spinal cord motor neuron cell fate specification
          spinal cord motor neuron differentiation
          visceral motor neuron differentiation
          trigeminal nerve development
          pituitary gland development
          neuron differentiation
          pancreas development
          axon regeneration
          retinal ganglion cell axon guidance
          positive regulation of insulin secretion
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 alpha production
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          negative regulation of intracellular estrogen receptor signaling pathway
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          negative regulation of neuron differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          neuron fate commitment
          neuron fate specification
          mesenchymal cell differentiation
          sensory system development
          peripheral nervous system neuron development
          peripheral nervous system neuron axonogenesis
          negative regulation of inflammatory response
          ventricular cardiac muscle tissue morphogenesis
          pharyngeal system development
          cardiac muscle cell myoblast differentiation
          innervation
          atrial septum morphogenesis
          cardiac cell fate determination
          cellular response to glucocorticoid stimulus
          positive regulation of granulocyte colony-stimulating factor production
          negative regulation of protein homodimerization activity
          negative regulation of canonical Wnt signaling pathway
          positive regulation of macrophage colony-stimulating factor production
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          enhancer sequence-specific DNA binding
          DNA binding
          chromatin binding
          protein binding
          zinc ion binding
          ligand-dependent nuclear receptor binding
          estrogen receptor binding
          bHLH transcription factor binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix