Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024268.2 | NP_001019439.1 | 28 | 2891 | ||
GenBank mRNA | AF276774.1 | - | 28 | 2921 | ||
BC093604.1 | - | 28 | 2875 | 7317599 | ||
BE104054.1 | - | 1 | 175 |
Gene Symbol: LOC100911727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759094.1 | XP_008757316.1 | 24 | 2560 |