Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024268.2 | NP_001019439.1 | 27 | 2779 | ||
GenBank mRNA | AF276774.1 | - | 27 | 2809 | ||
BC093604.1 | - | 27 | 2763 | 7317599 | ||
BE104054.1 | - | 2 | 287 |
Gene Symbol: LOC100911727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759094.1 | XP_008757316.1 | 23 | 2448 |