Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DEAF1 SIRNA Assays ›
          Gene Symbol:
          Deaf1
          Gene Name:
          DEAF1 transcription factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 214283787 - 214317466 on Build Rnor_6.0
          UniGene ID:
          Rn.11134
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS340309
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Deaf1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031801.1 NP_113989.1 10 1481
          XM_006230582.2 XP_006230644.1 10 2349
          XM_006230583.2 XP_006230645.1 10 2349
          XM_008760011.1 XP_008758233.1 10 1400
          XM_008760012.1 XP_008758234.1 10 1400
          XM_008760013.1 XP_008758235.1 10 1400
          XM_008760014.1 XP_008758236.1 10 2365
          XM_008760015.1 XP_008758237.1 5 993
          XM_008760016.1 XP_008758238.1 5 993
          GenBank mRNA AF055884.1 - 10 1481
          U59659.1 - 10 1227

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          neural tube closure
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          visual learning
          regulation of mammary gland epithelial cell proliferation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic skeletal system development
          DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-56f585df74-hnff6:80/100.66.130.62:80.
          git-commit: b2a1b94c9c7f5a2900588881f383e993a09fadb0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0