Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          UniGene ID:
          Rn.1868
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS340647
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012512.2 NP_036644.1 2 229
          GenBank mRNA FM032790.1 - 2 229
          FQ210051.1 - 2 242
          FQ210252.1 - 2 243
          FQ210804.1 - 2 241
          FQ210906.1 - 2 242
          FQ211296.1 - 2 242
          FQ211419.1 - 2 241
          FQ212410.1 - 2 243
          FQ217926.1 - 2 240
          FQ218141.1 - 2 242
          FQ218251.1 - 2 242
          FQ218298.1 - 2 241
          FQ219157.1 - 2 244
          FQ219455.1 - 791
          FQ220256.1 - 2 238
          FQ221287.1 - 2 238
          FQ221985.1 - 2 241
          FQ222352.1 - 2 239
          FQ225010.1 - 2 242
          FQ225051.1 - 2 246
          FQ225087.1 - 2 241
          FQ225088.1 - 2 243
          FQ225121.1 - 2 241
          FQ225141.1 - 2 241
          FQ225219.1 - 2 240
          FQ225292.1 - 2 242
          FQ225550.1 - 2 241
          FQ225647.1 - 2 241
          FQ225686.1 - 2 244
          FQ225690.1 - 2 383
          FQ225847.1 - 2 241
          FQ225912.1 - 2 241
          FQ226004.1 - 2 242
          FQ226258.1 - 2 244
          FQ226642.1 - 2 243
          FQ226723.1 - 2 241
          FQ227149.1 - 2 241
          FQ227291.1 - 2 247
          FQ227327.1 - 2 242
          FQ227454.1 - 2 242
          FQ227468.1 - 2 240
          FQ227656.1 - 2 242
          FQ227700.1 - 2 242
          FQ227950.1 - 2 240
          FQ227964.1 - 2 241
          FQ227995.1 - 2 237
          FQ228065.1 - 2 243
          FQ228188.1 - 2 242
          FQ229482.1 - 2 234
          FQ229718.1 - 2 241
          FQ230202.1 - 2 242
          FQ230402.1 - 2 242
          FQ230977.1 - 2 241
          FQ231135.1 - 2 242
          FQ231182.1 - 2 242
          FQ231367.1 - 2 241
          FQ231378.1 - 2 241
          FQ231524.1 - 2 241
          FQ231602.1 - 2 242
          FQ231718.1 - 2 241
          FQ231856.1 - 2 241
          FQ231894.1 - 2 241
          FQ232233.1 - 2 242
          FQ232294.1 - 2 242
          FQ232460.1 - 2 241
          FQ232758.1 - 2 241
          FQ232818.1 - 2 241
          FQ232819.1 - 2 241
          FQ232937.1 - 2 241
          FQ232943.1 - 2 243
          FQ232957.1 - 2 239
          FQ233220.1 - 2 242
          FQ233240.1 - 2 242
          FQ233290.1 - 2 241
          FQ233447.1 - 2 242
          FQ233505.1 - 2 241
          FQ233544.1 - 2 241
          FQ233567.1 - 2 246
          FQ233757.1 - 2 241
          FQ233797.1 - 2 240
          FQ233849.1 - 2 241
          FQ234348.1 - 2 242
          FQ234349.1 - 2 241
          FQ234524.1 - 2 239
          FQ234581.1 - 2 239
          FQ234840.1 - 2 241
          FQ234885.1 - 2 241
          FQ235149.1 - 2 239
          FQ235229.1 - 2 241
          X16956.1 - 1 18
          Y00441.1 - 2 216
          Y08531.1 - 2 221

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          T cell differentiation in thymus
          protein refolding
          response to drug
          response to cadmium ion
          iron ion homeostasis
          molecular_function
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-bff4f7d6d-tmrcx:80/100.66.135.11:80.
          git-commit: 683fd26f943b2dde6a08880618459b58d20c3896
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline