Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012512.2 | NP_036644.1 | 2 | 330 | ||
GenBank mRNA | FM032790.1 | - | 2 | 330 | ||
FQ210051.1 | - | 2 | 343 | |||
FQ210252.1 | - | 2 | 344 | |||
FQ210804.1 | - | 2 | 342 | |||
FQ210906.1 | - | 2 | 343 | |||
FQ211296.1 | - | 2 | 343 | |||
FQ211419.1 | - | 2 | 342 | |||
FQ212410.1 | - | 2 | 344 | |||
FQ217926.1 | - | 2 | 341 | |||
FQ218141.1 | - | 2 | 343 | |||
FQ218251.1 | - | 2 | 343 | |||
FQ218298.1 | - | 2 | 342 | |||
FQ219157.1 | - | 2 | 345 | |||
FQ219455.1 | - | 892 | ||||
FQ220256.1 | - | 2 | 339 | |||
FQ221268.1 | - | 2 | 343 | |||
FQ221287.1 | - | 2 | 339 | |||
FQ221985.1 | - | 2 | 342 | |||
FQ222352.1 | - | 2 | 340 | |||
FQ225010.1 | - | 2 | 343 | |||
FQ225051.1 | - | 2 | 347 | |||
FQ225087.1 | - | 2 | 342 | |||
FQ225088.1 | - | 2 | 344 | |||
FQ225121.1 | - | 2 | 342 | |||
FQ225141.1 | - | 2 | 342 | |||
FQ225219.1 | - | 2 | 341 | |||
FQ225292.1 | - | 2 | 343 | |||
FQ225550.1 | - | 2 | 342 | |||
FQ225647.1 | - | 2 | 342 | |||
FQ225686.1 | - | 2 | 345 | |||
FQ225690.1 | - | 2 | 484 | |||
FQ225847.1 | - | 2 | 342 | |||
FQ226004.1 | - | 2 | 343 | |||
FQ226258.1 | - | 2 | 345 | |||
FQ226642.1 | - | 2 | 344 | |||
FQ227149.1 | - | 2 | 342 | |||
FQ227291.1 | - | 2 | 348 | |||
FQ227327.1 | - | 2 | 343 | |||
FQ227454.1 | - | 2 | 343 | |||
FQ227468.1 | - | 2 | 341 | |||
FQ227656.1 | - | 2 | 343 | |||
FQ227950.1 | - | 2 | 341 | |||
FQ227964.1 | - | 2 | 342 | |||
FQ227995.1 | - | 2 | 338 | |||
FQ228065.1 | - | 2 | 344 | |||
FQ228188.1 | - | 2 | 343 | |||
FQ228427.1 | - | 2 | 344 | |||
FQ229482.1 | - | 2 | 335 | |||
FQ229718.1 | - | 2 | 342 | |||
FQ230202.1 | - | 2 | 343 | |||
FQ230402.1 | - | 2 | 343 | |||
FQ230977.1 | - | 2 | 342 | |||
FQ231135.1 | - | 2 | 343 | |||
FQ231182.1 | - | 2 | 343 | |||
FQ231367.1 | - | 2 | 342 | |||
FQ231378.1 | - | 2 | 342 | |||
FQ231524.1 | - | 2 | 342 | |||
FQ231602.1 | - | 2 | 343 | |||
FQ231718.1 | - | 2 | 342 | |||
FQ231856.1 | - | 2 | 342 | |||
FQ232233.1 | - | 2 | 343 | |||
FQ232294.1 | - | 2 | 343 | |||
FQ232460.1 | - | 2 | 342 | |||
FQ232758.1 | - | 2 | 342 | |||
FQ232818.1 | - | 2 | 342 | |||
FQ232819.1 | - | 2 | 342 | |||
FQ232937.1 | - | 2 | 342 | |||
FQ232943.1 | - | 2 | 344 | |||
FQ232957.1 | - | 2 | 340 | |||
FQ233220.1 | - | 2 | 343 | |||
FQ233240.1 | - | 2 | 343 | |||
FQ233290.1 | - | 2 | 342 | |||
FQ233447.1 | - | 2 | 343 | |||
FQ233505.1 | - | 2 | 342 | |||
FQ233544.1 | - | 2 | 342 | |||
FQ233567.1 | - | 2 | 347 | |||
FQ233757.1 | - | 2 | 342 | |||
FQ233797.1 | - | 2 | 341 | |||
FQ233849.1 | - | 2 | 342 | |||
FQ234348.1 | - | 2 | 343 | |||
FQ234349.1 | - | 2 | 342 | |||
FQ234581.1 | - | 2 | 340 | |||
FQ234840.1 | - | 2 | 342 | |||
FQ234885.1 | - | 2 | 342 | |||
FQ235105.1 | - | 2 | 342 | |||
FQ235149.1 | - | 2 | 340 | |||
FQ235229.1 | - | 2 | 342 | |||
X16956.1 | - | 1 | 119 | |||
Y00441.1 | - | 2 | 317 | |||
Y08531.1 | - | 2 | 322 |