Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024355.1 | NP_077331.1 | 9 | 2393 | ||
GenBank mRNA | AF017757.1 | - | 9 | 2393 |