Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106236.1 | NP_001099706.1 | 2, 3 | 386 | ||
XM_008759117.1 | XP_008757339.1 | 2, 3 | 307 | |||
GenBank mRNA | BC168943.1 | - | 2, 3 | 290 | 7322093 | |
FQ228106.1 | - | 2, 3 | 270 | |||
FQ232598.1 | - | 2, 3 | 333 |