Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTA1 SIRNA Assays ›
          Gene Symbol:
          Acta1
          Gene Name:
          actin, alpha 1, skeletal muscle
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56674072 - 56677084 on Build Rnor_6.0
          UniGene ID:
          Rn.82732
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS341452
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Acta1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019212.2 NP_062085.1 6 936
          GenBank mRNA BC061974.1 - 6 936 5623909
          FQ214585.1 - 6 947
          FQ214598.1 - 6 947
          FQ214600.1 - 6 946
          FQ214603.1 - 6 949
          FQ214610.1 - 4 490
          FQ214623.1 - 6 944
          FQ214640.1 - 6 945
          FQ214659.1 - 6 947
          FQ214676.1 - 6 946
          FQ214680.1 - 6 946
          FQ214697.1 - 6 951
          FQ214734.1 - 6 946
          FQ214743.1 - 6 945
          FQ214756.1 - 6 947
          FQ214761.1 - 6 944
          FQ214779.1 - 6 944
          FQ214785.1 - 6 944
          FQ214786.1 - 6 948
          FQ214807.1 - 6 945
          FQ214831.1 - 6 946
          FQ214832.1 - 6 944
          FQ214839.1 - 6 948
          FQ214841.1 - 6 945
          FQ214842.1 - 6 946
          FQ214858.1 - 6 945
          FQ214859.1 - 6 945
          FQ214866.1 - 6 947
          FQ214883.1 - 6 944
          FQ214904.1 - 6 945
          FQ214905.1 - 6 946
          FQ214916.1 - 6 945
          FQ214921.1 - 6 946
          FQ214936.1 - 6 946
          FQ214948.1 - 6 945
          FQ214949.1 - 6 946
          FQ214957.1 - 6 945
          FQ214969.1 - 6 945
          FQ214975.1 - 6 948
          FQ214976.1 - 6 945
          FQ214978.1 - 6 945
          FQ214987.1 - 6 945
          FQ214990.1 - 6 945
          FQ214993.1 - 6 945
          FQ215012.1 - 6 946
          FQ215025.1 - 6 947
          FQ215038.1 - 6 944
          FQ215044.1 - 6 945
          FQ215089.1 - 6 946
          FQ215112.1 - 6 945
          FQ215128.1 - 6 947
          FQ215136.1 - 6 945
          FQ215153.1 - 6 945
          FQ215158.1 - 6 943
          FQ215173.1 - 6 946
          FQ215185.1 - 6 945
          FQ215204.1 - 6 943
          FQ215212.1 - 6 947
          FQ215225.1 - 6 946
          FQ215241.1 - 6 946
          FQ215249.1 - 6 946
          FQ215253.1 - 6 946
          FQ215256.1 - 6 945
          FQ215275.1 - 6 945
          FQ215277.1 - 6 946
          FQ215280.1 - 6 946
          FQ215282.1 - 6 945
          FQ215298.1 - 6 946
          FQ215324.1 - 6 945
          FQ215342.1 - 6 945
          FQ215346.1 - 6 945
          FQ215366.1 - 6 946
          FQ215381.1 - 6 946
          FQ215393.1 - 6 945
          FQ215396.1 - 6 945
          FQ215398.1 - 6 945
          FQ215402.1 - 6 945
          FQ215403.1 - 6 946
          FQ215405.1 - 6 944
          FQ215414.1 - 6 947
          FQ215431.1 - 6 946
          FQ215433.1 - 6 944
          FQ215434.1 - 6 947
          FQ215446.1 - 6 945
          FQ215449.1 - 6 948
          FQ215454.1 - 6 946
          FQ215464.1 - 6 945
          FQ215465.1 - 6 945
          FQ215470.1 - 6 945
          FQ215474.1 - 6 946
          FQ215483.1 - 6 945
          FQ215488.1 - 6 945
          FQ215518.1 - 6 954
          FQ215523.1 - 6 945
          FQ215537.1 - 6 947
          FQ215554.1 - 6 945
          FQ215567.1 - 6 947
          FQ215572.1 - 6 946
          FQ215593.1 - 6 945
          FQ215618.1 - 6 946
          FQ215631.1 - 4 559
          FQ215655.1 - 6 943
          FQ215659.1 - 6 944
          FQ215673.1 - 6 945
          FQ215674.1 - 6 945
          FQ215690.1 - 6 946
          FQ215692.1 - 6 946
          FQ215790.1 - 6 947
          FQ215799.1 - 6 944
          FQ215809.1 - 6 945
          FQ215813.1 - 6 945
          FQ215835.1 - 6 945
          FQ215839.1 - 6 945
          FQ215840.1 - 6 946
          FQ215851.1 - 6 945
          FQ215852.1 - 6 943
          FQ215864.1 - 6 947
          FQ215866.1 - 6 946
          FQ215888.1 - 6 944
          FQ215893.1 - 6 948
          FQ215896.1 - 6 945
          FQ215950.1 - 6 947
          FQ215964.1 - 6 945
          FQ215972.1 - 6 947
          FQ216016.1 - 6 945
          FQ216035.1 - 6 945
          FQ216043.1 - 6 945
          FQ216050.1 - 6 945
          FQ216066.1 - 6 943
          FQ216138.1 - 6 946
          FQ216139.1 - 6 945
          FQ216168.1 - 6 944
          FQ216196.1 - 6 946
          FQ216224.1 - 6 946
          FQ216278.1 - 6 949
          FQ216287.1 - 6 945
          FQ216291.1 - 6 946
          FQ216293.1 - 6 946
          FQ216305.1 - 6 946
          FQ216333.1 - 6 946
          FQ216350.1 - 6 945
          FQ216352.1 - 6 945
          FQ216360.1 - 6 946
          FQ216369.1 - 6 948
          FQ216403.1 - 6 945
          FQ216430.1 - 6 946
          FQ216509.1 - 6 943
          FQ216586.1 - 6 945
          FQ216603.1 - 6 947
          FQ216606.1 - 6 945
          FQ216607.1 - 6 941
          FQ216610.1 - 6 943
          FQ216620.1 - 6 946
          FQ216635.1 - 6 943
          FQ216639.1 - 6 945
          FQ216644.1 - 6 946
          FQ216664.1 - 6 943
          FQ216676.1 - 6 945
          FQ216692.1 - 6 946
          FQ216703.1 - 6 945
          FQ216713.1 - 6 945
          FQ216723.1 - 6 946
          FQ216725.1 - 6 941
          FQ216735.1 - 2 185
          FQ216739.1 - 6 945
          FQ216762.1 - 6 943
          FQ216767.1 - 6 945
          FQ216776.1 - 6 946
          FQ216777.1 - 6 945
          FQ216779.1 - 6 947
          FQ216793.1 - 6 945
          FQ216799.1 - 6 943
          FQ216801.1 - 6 945
          FQ216819.1 - 6 943
          FQ216916.1 - 6 947
          FQ216918.1 - 4 588
          FQ216961.1 - 6 946
          FQ216994.1 - 6 945
          FQ216999.1 - 2 144
          FQ217061.1 - 6 947
          FQ217073.1 - 6 948
          FQ217091.1 - 6 945
          FQ217114.1 - 6 947
          FQ217167.1 - 6 945
          FQ217173.1 - 6 945
          FQ217188.1 - 6 945
          FQ217194.1 - 6 945
          FQ217266.1 - 1 72
          FQ217273.1 - 6 944
          FQ217280.1 - 6 1065
          FQ217327.1 - 6 946
          FQ217329.1 - 6 945
          FQ217347.1 - 6 944
          FQ217351.1 - 1 28
          FQ217353.1 - 6 945
          FQ217367.1 - 6 945
          FQ217397.1 - 6 945
          FQ217441.1 - 6 945
          FQ217443.1 - 6 947
          FQ217475.1 - 6 946
          FQ217525.1 - 6 945
          FQ217535.1 - 6 945
          FQ217572.1 - 6 945
          FQ217593.1 - 6 945
          FQ217602.1 - 6 947
          FQ217615.1 - 6 945
          FQ217629.1 - 6 947
          FQ217641.1 - 6 942
          FQ217668.1 - 2 194
          FQ217762.1 - 6 945
          FQ217855.1 - 1 20
          FQ217884.1 - 6 944
          FQ217905.1 - 6 945
          FQ217908.1 - 6 947
          FQ217937.1 - 6 945
          FQ217969.1 - 6 947
          FQ218005.1 - 3 345
          FQ218019.1 - 6 946
          FQ218034.1 - 6 945
          FQ223563.1 - 6 946
          FQ223686.1 - 6 945
          FQ223702.1 - 6 946
          FQ223727.1 - 4 643
          FQ223778.1 - 4 459
          FQ223898.1 - 6 946
          FQ223914.1 - 6 945
          FQ223926.1 - 6 945
          FQ223929.1 - 6 944
          FQ224009.1 - 6 945
          FQ224082.1 - 6 945
          FQ224141.1 - 6 946
          FQ224359.1 - 6 946
          FQ224377.1 - 6 945
          FQ224552.1 - 6 946
          FQ224691.1 - 6 943
          FQ224777.1 - 6 946
          FQ224840.1 - 6 943
          FQ224842.1 - 6 945
          FQ224862.1 - 6 945
          FQ224869.1 - 6 946
          FQ224886.1 - 6 944
          FQ224890.1 - 6 943
          FQ224902.1 - 6 946
          FQ224929.1 - 6 943
          FQ224936.1 - 6 946
          FQ224942.1 - 6 948
          FQ224946.1 - 4 473
          FQ224948.1 - 6 950
          FQ224962.1 - 6 947
          FQ224971.1 - 6 945

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001100, AK096902, BX648545, AK300697, AY280960, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001272041, NM_009606, M12866, BC014877, X03766, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          response to mechanical stimulus
          response to extracellular stimulus
          response to lithium ion
          cell growth
          skeletal muscle thin filament assembly
          skeletal muscle fiber adaptation
          response to steroid hormone
          skeletal muscle fiber development
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0