Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100361056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252979.2 | XP_006253041.1 | 1 | 888 |
Gene Symbol: Tmem60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191610.1 | NP_001178539.1 | 2 | 563 |