Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phykpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128196.1 | NP_001121668.1 | 10 | 1108 | ||
XM_008767645.1 | XP_008765867.1 | 10 | 1906 | |||
XM_008767647.1 | XP_008765869.1 | 10 | 1169 | |||
XM_008767648.1 | XP_008765870.1 | 10 | 1906 | |||
XM_008767649.1 | XP_008765871.1 | 10 | 1906 | |||
XM_008767650.1 | XP_008765872.1 | 10 | 1906 | |||
XM_008767651.1 | XP_008765873.1 | 10 | 1906 | |||
XM_008767652.1 | XP_008765874.1 | 10 | 1906 | |||
GenBank mRNA | BC079163.1 | - | 10 | 1121 | ||
BC158766.1 | - | 10 | 1108 | 7114211 | ||
FQ209628.1 | - | 2 | 137 |
Gene Symbol: LOC103690164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246210.2 | XP_006246272.1 | 8 | 1076 | ||
XM_008767733.1 | XP_008765955.1 | 10 | 1906 | |||
XM_008767734.1 | XP_008765956.1 | 1906 | ||||
XM_008767735.1 | XP_008765957.1 | 10 | 1906 | |||
XM_008767736.1 | XP_008765958.1 | 10 | 1906 | |||
XM_008767737.1 | XP_008765959.1 | 1906 | ||||
XM_008767738.1 | XP_008765960.1 | 10 | 1906 |