Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153814.2 | NP_722521.1 | 19 | 4077 | ||
XM_008767532.1 | XP_008765754.1 | 18 | 8456 | |||
XM_008767533.1 | XP_008765755.1 | 18 | 8456 | |||
XM_008767534.1 | XP_008765756.1 | 18 | 8456 | |||
XM_008767535.1 | XP_008765757.1 | 18 | 8456 | |||
XM_008767536.1 | XP_008765758.1 | 17 | 3675 | |||
GenBank mRNA | AF290213.2 | - | 19 | 4080 |