Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC498276 SIRNA Assays ›
          Gene Symbol:
          LOC498276 Fcgr2a LOC100911825 LOC103693683 LOC103694908
          Gene Name:
          Fc gamma receptor II beta
          Fc fragment of IgG, low affinity IIa, receptor
          low affinity immunoglobulin gamma Fc region receptor III-like
          low affinity immunoglobulin gamma Fc region receptor III
          low affinity immunoglobulin gamma Fc region receptor III-like
          Gene Aliases:
          -
          Fcgr3, Fcgr3a
          -
          -
          -
          Chromosome Location:
          Chr.13: 91168972 - 91181954 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 91146879 - 91163694 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 91219838 - 91237277 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 89420835 - 89440644 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906965: 601 - 13578 on Build Rnor_6.0
          UniGene ID:
          Rn.6050
          Rn.154415
          -
          Rn.6050
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID RSS350362
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC498276
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135992.1 NP_001129464.1 762
          XM_008769748.1 XP_008767970.1 723
          XM_008769749.1 XP_008767971.1 723
          XM_008769750.1 XP_008767972.1 847
          GenBank mRNA BC159405.1 - 762 7459664
          Gene Symbol: Fcgr2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053843.2 NP_446295.2 6 902
          XM_006250303.2 XP_006250365.1 6 955
          XM_006250305.2 XP_006250367.1 6 1054
          GenBank mRNA BC060534.1 - 679 6886727
          L08446.1 - 6 902
          M32062.1 - 715
          M64368.1 - 695
          M64369.1 - 695
          M64370.1 - 695
          Gene Symbol: LOC100911825
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006250308.2 XP_006250370.1 5 723
          XM_008769774.1 XP_008767996.1 5 723
          XM_008769775.1 XP_008767997.1 6 948
          XM_008769776.1 XP_008767998.1 6 756
          XM_008769777.1 XP_008767999.1 4 702
          Gene Symbol: LOC103693683
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008769767.1 XP_008767989.1 6 744
          XM_008769768.1 XP_008767990.1 6 949
          XM_008769769.1 XP_008767991.1 6 1536
          XM_008769770.1 XP_008767992.1 6 1518
          XM_008769771.1 XP_008767993.1 5 725
          XM_008769772.1 XP_008767994.1 6 1129
          XM_008769773.1 XP_008767995.1 5 707
          Gene Symbol: LOC103694908
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773904.1 XP_008772126.1 723
          XM_008773905.1 XP_008772127.1 724
          XM_008773906.1 XP_008772128.1 851

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          nerve development
          Fc-gamma receptor signaling pathway
          IgG receptor activity
          IgG binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix