Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM3 SIRNA Assays ›
Gene Symbol:
Tpm3
Gene Name:
tropomyosin 3
Gene Aliases:
TM30nm, Tpm5
Chromosome Location:
Chr.2: 189423539 - 189451345 on Build Rnor_6.0
UniGene ID:
Rn.37575
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS350571
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301285.1
NP_001288214.1
3, 4
466
NM_001301286.1
NP_001288215.1
2, 3
290
NM_057208.2
NP_476556.2
2, 3
290
NM_173111.1
NP_775134.1
2, 3
332
XM_006232568.2
XP_006232630.1
3, 4
486
XM_006232569.2
XP_006232631.1
3, 4
486
XM_006232570.2
XP_006232632.1
3, 4
486
XM_006232571.2
XP_006232633.1
3, 4
486
XM_006232572.2
XP_006232634.1
3, 4
486
XM_006232573.2
XP_006232635.1
3, 4
486
XM_006232575.2
XP_006232637.1
3, 4
486
XM_006232576.2
XP_006232638.1
3, 4
486
XM_006232577.2
XP_006232639.1
3, 4
486
XM_006232580.2
XP_006232642.1
2, 3
390
XM_006232581.2
XP_006232643.1
2, 3
391
XM_006232583.2
XP_006232645.1
2, 3
394
XM_006232584.2
XP_006232646.1
2, 3
392
XM_008761124.1
XP_008759346.1
2, 3
390
XM_008761125.1
XP_008759347.1
2, 3
491
GenBank mRNA
FQ214881.1
-
3, 4
464
FQ215580.1
-
3, 4
466
FQ215752.1
-
3, 4
466
FQ216429.1
-
3, 4
465
FQ228755.1
-
2, 3
340
L24775.1
-
2, 3
290
L24776.1
-
290
L24777.1
-
2, 3
290
S82383.1
-
2, 3
309
X72859.1
-
2, 3
332
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001271764, NM_001253738, NM_001253740, NM_022314, AK166836, and others.
Gene Ontology Categories:
Function(s)
Process(es)
brain development
molecular_function
actin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0