Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOX9 SIRNA Assays ›
          Gene Symbol:
          Sox9
          Gene Name:
          SRY (sex determining region Y)-box 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 101288354 - 101294030 on Build Rnor_6.0
          UniGene ID:
          Rn.95086
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS350590
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sox9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003750950.3 XP_003750998.1 1 490

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_011448, BC024958, AK031516, BC023796, BC034264, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          cartilage condensation
          ossification
          branching involved in ureteric bud morphogenesis
          cell fate specification
          epithelial to mesenchymal transition
          tissue homeostasis
          positive regulation of protein phosphorylation
          hair follicle development
          positive regulation of mesenchymal cell proliferation
          chondrocyte development
          negative regulation of immune system process
          heart valve development
          heart valve morphogenesis
          heart valve formation
          endocardial cushion morphogenesis
          chondrocyte differentiation involved in endochondral bone morphogenesis
          chondrocyte hypertrophy
          nucleosome assembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          protein complex assembly
          protein phosphorylation
          cytoskeleton organization
          signal transduction
          epidermal growth factor receptor signaling pathway
          spermatogenesis
          heart development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          male gonad development
          regulation of cell cycle process
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial cell migration
          neural crest cell development
          positive regulation of phosphatidylinositol 3-kinase signaling
          single organismal cell-cell adhesion
          male germ-line sex determination
          cAMP-mediated signaling
          cell differentiation
          regulation of cell adhesion
          extracellular matrix organization
          male sex determination
          negative regulation of ossification
          negative regulation of bone mineralization
          prostate gland development
          negative regulation of epithelial cell differentiation
          positive regulation of epithelial cell differentiation
          mammary gland development
          notochord development
          otic vesicle formation
          endocrine pancreas development
          negative regulation of chondrocyte differentiation
          positive regulation of chondrocyte differentiation
          lacrimal gland development
          protein localization to nucleus
          somatic stem cell population maintenance
          intrahepatic bile duct development
          regulation of cell proliferation
          regulation of apoptotic process
          negative regulation of apoptotic process
          protein kinase B signaling
          cell fate commitment
          regulation of cell differentiation
          negative regulation of myoblast differentiation
          positive regulation of protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of photoreceptor cell differentiation
          oligodendrocyte differentiation
          homeostasis of number of cells within a tissue
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          cartilage development
          Sertoli cell differentiation
          Sertoli cell development
          astrocyte fate commitment
          retina development in camera-type eye
          limb bud formation
          retinal rod cell differentiation
          endochondral bone morphogenesis
          epithelial tube branching involved in lung morphogenesis
          lung epithelial cell differentiation
          prostate gland morphogenesis
          epithelial cell proliferation involved in prostatic bud elongation
          bronchus cartilage development
          trachea cartilage development
          intestinal epithelial structure maintenance
          regulation of cell proliferation involved in tissue homeostasis
          positive regulation of cartilage development
          regulation of branching involved in lung morphogenesis
          morphogenesis of a branching epithelium
          lung smooth muscle development
          negative regulation of biomineral tissue development
          ERK1 and ERK2 cascade
          Harderian gland development
          cellular response to mechanical stimulus
          cellular response to retinoic acid
          cellular response to interleukin-1
          cellular response to epidermal growth factor stimulus
          cellular response to heparin
          cellular response to transforming growth factor beta stimulus
          renal vesicle induction
          ureter development
          ureter urothelium development
          ureter smooth muscle cell differentiation
          ureter morphogenesis
          metanephric nephron tubule formation
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          positive regulation of kidney development
          positive regulation of branching involved in ureteric bud morphogenesis
          positive regulation of extracellular matrix assembly
          positive regulation of male gonad development
          positive regulation of cell proliferation involved in heart morphogenesis
          positive regulation of mesenchymal stem cell differentiation
          regulation of epithelial cell proliferation involved in lung morphogenesis
          negative regulation of mesenchymal cell apoptotic process
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein kinase activity
          protein binding
          beta-catenin binding
          protein kinase A catalytic subunit binding
          enhancer binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          pre-mRNA intronic binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0