Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017014.1 | NP_058710.1 | 8 | 675 | ||
GenBank mRNA | BC063172.1 | - | 8 | 715 | 6920688 | |
FJ179401.1 | - | 8 | 639 | |||
FQ211067.1 | - | 8 | 674 | |||
FQ212742.1 | - | 8 | 668 | |||
FQ213204.1 | - | 8 | 668 | |||
FQ213696.1 | - | 8 | 862 | |||
FQ214055.1 | - | 8 | 774 | |||
FQ217092.1 | - | 8 | 741 | |||
FQ218381.1 | - | 8 | 675 | |||
FQ218789.1 | - | 8 | 674 | |||
FQ218795.1 | - | 8 | 711 | |||
FQ229941.1 | - | 8 | 702 | |||
J02810.1 | - | 8 | 675 | |||
M11719.1 | - | 8 | 676 | |||
M28241.1 | - | 8 | 735 | |||
X04229.1 | - | 8 | 675 |