Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173095.3 | NP_775118.1 | 1 | 1089 | ||
GenBank mRNA | M26161.1 | - | 1 | 1089 | ||
X12589.1 | - | 1302 |