Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012607.2 | NP_036739.2 | 3 | 1243 | ||
XM_006251180.2 | XP_006251242.1 | 3 | 1269 | |||
XM_006251181.2 | XP_006251243.1 | 3 | 1269 | |||
XM_006251183.2 | XP_006251245.1 | 3 | 1269 | |||
XM_006251184.2 | XP_006251246.1 | 3 | 1269 | |||
XM_006251185.2 | XP_006251247.1 | 3 | 1269 | |||
XM_006251186.2 | XP_006251248.1 | 3 | 1269 | |||
XM_006251187.2 | XP_006251249.1 | 3 | 1269 | |||
XM_008770256.1 | XP_008768478.1 | 3 | 1269 | |||
XM_008770257.1 | XP_008768479.1 | 3 | 1269 | |||
GenBank mRNA | AF031879.1 | - | 1242 | |||
CB582861.1 | - | 2 | 151 | |||
M21964.1 | - | 3 | 384 |