Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076553.2 | NP_001070021.1 | 5, 6 | 544 | ||
NM_001271158.1 | NP_001258087.1 | 4, 5 | 379 | |||
NM_012627.3 | NP_036759.2 | 4, 5 | 439 | |||
XM_006256511.2 | XP_006256573.1 | 5, 6 | 1163 | |||
XM_006256512.2 | XP_006256574.1 | 6, 7 | 666 | |||
XM_006256513.2 | XP_006256575.1 | 5, 6 | 521 | |||
XM_006256515.2 | XP_006256577.1 | 5, 6 | 1086 | |||
XM_008772976.1 | XP_008771198.1 | 6, 7 | 803 | |||
XM_008772977.1 | XP_008771199.1 | 6, 7 | 1516 | |||
XM_008772978.1 | XP_008771200.1 | 5, 6 | 1253 | |||
GenBank mRNA | AF413572.1 | - | 5, 6 | 527 | ||
BC078880.1 | - | 4, 5 | 379 | 7111554 | ||
FQ231722.1 | - | 4, 5 | 435 | |||
M64092.1 | - | 4, 5 | 396 |