Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019381.2 | NP_062254.2 | 4 | 293 | ||
XM_006257323.2 | XP_006257385.1 | 4 | 435 | |||
XM_008765670.1 | XP_008763892.1 | 5 | 393 | |||
GenBank mRNA | AY321320.1 | - | 6 | 431 | ||
AY325130.1 | - | 6 | 431 | |||
AY325243.1 | - | 6 | 431 | |||
BC058478.1 | - | 4 | 299 | 6888036 | ||
FQ217052.1 | - | 4 | 292 | |||
FQ218664.1 | - | 4 | 295 | |||
FQ226858.1 | - | 4 | 322 | |||
FQ229652.1 | - | 4 | 294 | |||
FQ230608.1 | - | 4 | 322 | |||
FQ231817.1 | - | 4 | 300 | |||
X75855.1 | - | 4 | 325 | |||
X75856.1 | - | 3 | 209 |