Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013110.1 | NP_001013128.1 | 16 | 1966 | ||
GenBank mRNA | AF476964.1 | - | 17 | 1947 | ||
AY327504.1 | - | 16 | 1935 | |||
BC087021.1 | - | 16 | 1966 | 7132382 | ||
D38380.1 | - | 16 | 1965 | |||
FQ209414.1 | - | 9 | 930 | |||
FQ209524.1 | - | 9 | 1026 | |||
FQ209552.1 | - | 7 | 745 | |||
FQ209673.1 | - | 10 | 1133 | |||
FQ209674.1 | - | 10 | 1191 | |||
FQ209794.1 | - | 8 | 818 | |||
FQ210028.1 | - | 5 | 551 | |||
FQ210115.1 | - | 4 | 446 | |||
FQ210327.1 | - | 4 | 422 | |||
FQ210637.1 | - | 4 | 364 | |||
FQ210963.1 | - | 5 | 490 | |||
FQ211385.1 | - | 7 | 742 | |||
FQ211460.1 | - | 2 | 248 | |||
FQ213988.1 | - | 6 | 651 | |||
FQ214362.1 | - | 9 | 1043 | |||
FQ218210.1 | - | 8 | 796 | |||
FQ219264.1 | - | 10 | 1069 | |||
FQ219315.1 | - | 5 | 607 | |||
FQ219349.1 | - | 8 | 799 | |||
FQ219354.1 | - | 2 | 557 | |||
FQ219664.1 | - | 5 | 460 | |||
M27966.1 | - | 4 | 375 | |||
X77158.1 | - | 16 | 1943 |
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY325214.1 | - | 1933 | |||
AY325230.1 | - | 1936 | ||||
AY325241.1 | - | 1933 |