Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS351741
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
5
619
NM_001034069.1
NP_001029241.1
5
619
NM_001034070.1
NP_001029242.1
5
619
NM_001034071.1
NP_001029243.1
5
619
NM_001034072.1
NP_001029244.1
5
619
NM_001034073.1
NP_001029245.1
4
556
NM_001034074.1
NP_001029246.1
4
556
NM_001034075.1
NP_001029247.1
4
556
NM_001301336.1
NP_001288265.1
5
619
NM_001301342.1
NP_001288271.1
5
619
NM_001301736.1
NP_001288665.1
4
556
NM_019131.2
NP_062004.1
4
556
XM_006243386.2
XP_006243448.1
5
611
XM_006243388.2
XP_006243450.1
5
611
XM_006243389.2
XP_006243451.1
5
611
XM_006243390.2
XP_006243452.1
5
611
XM_006243393.1
XP_006243455.1
4
554
XM_006243394.1
4
572
XM_006243395.2
XP_006243457.1
4
563
XM_008766207.1
XP_008764429.1
5
611
XM_008766208.1
XP_008764430.1
4
554
GenBank mRNA
AF372215.1
-
4
405
AF372216.1
-
5
518
AF372217.1
-
5
524
BC078780.1
-
4
523
7106337
CB585296.1
-
4
559
CF108680.1
-
5
567
CK483405.1
-
5
573
FQ074026.1
-
5
677
FQ214619.1
-
5
594
FQ214809.1
-
5
588
FQ214835.1
-
5
588
FQ214864.1
-
5
592
FQ214868.1
-
5
588
FQ214869.1
-
5
594
FQ214894.1
-
5
594
FQ214902.1
-
5
588
FQ214911.1
-
5
590
FQ214944.1
-
5
594
FQ214946.1
-
5
594
FQ214991.1
-
5
588
FQ215107.1
-
5
588
FQ215186.1
-
5
594
FQ215188.1
-
5
588
FQ215206.1
-
5
623
FQ215226.1
-
5
594
FQ215459.1
-
5
590
FQ215512.1
-
5
594
FQ215522.1
-
5
665
FQ215524.1
-
5
590
FQ215538.1
-
5
594
FQ215551.1
-
5
589
FQ215698.1
-
5
588
FQ215703.1
-
5
588
FQ215724.1
-
5
675
FQ215746.1
-
5
590
FQ215761.1
-
5
588
FQ215768.1
-
5
588
FQ215803.1
-
5
594
FQ215808.1
-
5
590
FQ215815.1
-
5
588
FQ215919.1
-
5
596
FQ215944.1
-
5
595
FQ215969.1
-
5
588
FQ215995.1
-
5
590
FQ216014.1
-
5
672
FQ216018.1
-
5
594
FQ216052.1
-
5
588
FQ216118.1
-
5
672
FQ216145.1
-
5
594
FQ216160.1
-
5
587
FQ216319.1
-
5
590
FQ216340.1
-
5
594
FQ216368.1
-
5
590
FQ216400.1
-
5
590
FQ216494.1
-
5
588
FQ216552.1
-
5
594
FQ216557.1
-
5
594
FQ216569.1
-
683
FQ216615.1
-
5
503
FQ216637.1
-
5
590
FQ216683.1
-
5
590
FQ216697.1
-
5
590
FQ216747.1
-
5
590
FQ217468.1
-
4
339
FQ217503.1
-
5
594
FQ217616.1
-
3
156
FQ217637.1
-
5
594
FQ217808.1
-
5
590
FQ223706.1
-
5
590
FQ223993.1
-
5
592
FQ224036.1
-
5
505
FQ224207.1
-
5
588
FQ224838.1
-
5
594
FQ224943.1
-
5
592
FQ224961.1
-
5
590
M34134.1
-
4
541
M34135.1
-
5
553
M34136.1
-
3
271
M60666.1
-
5
566
M60667.1
-
5
566
M60668.1
-
4
503
M60669.1
-
4
503
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0