Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DOCK9 SIRNA Assays ›
Gene Symbol:
Dock9
Gene Name:
dedicator of cytokinesis 9
Gene Aliases:
Trg
Chromosome Location:
Chr.15: 107945664 - 108210985 on Build Rnor_6.0
UniGene ID:
Rn.10431
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS352404
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dock9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105759.2
NP_001099229.2
26
2943
XM_006252481.2
XP_006252543.1
26
3054
XM_006252488.2
XP_006252550.1
26
3054
XM_006252490.2
XP_006252552.1
26
3054
XM_006252493.2
XP_006252555.1
26
3509
XM_006252496.2
XP_006252558.1
26
3078
XM_006252499.2
XP_006252561.1
26
3053
XM_006252501.2
XP_006252563.1
26
3053
XM_006252502.2
XP_006252564.1
26
3053
XM_006252503.2
XP_006252565.1
26
3102
XM_006252505.2
XP_006252567.1
26
3078
XM_008770951.1
XP_008769173.1
26
3054
XM_008770952.1
XP_008769174.1
26
3054
XM_008770953.1
XP_008769175.1
26
3054
XM_008770954.1
XP_008769176.1
26
3054
XM_008770955.1
XP_008769177.1
26
3079
XM_008770956.1
XP_008769178.1
26
3079
XM_008770957.1
XP_008769179.1
26
3054
XM_008770958.1
XP_008769180.1
26
3054
XM_008770959.1
XP_008769181.1
26
3054
XM_008770960.1
XP_008769182.1
26
3054
XM_008770961.1
XP_008769183.1
26
3054
XM_008770962.1
XP_008769184.1
26
3054
XM_008770963.1
XP_008769185.1
26
3054
XM_008770964.1
XP_008769186.1
26
3119
XM_008770965.1
XP_008769187.1
26
3054
XM_008770966.1
XP_008769188.1
26
3054
XM_008770967.1
XP_008769189.1
26
3053
XM_008770968.1
XP_008769190.1
26
3514
XM_008770969.1
XP_008769191.1
26
3515
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
Rho GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0