Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DOCK9 SIRNA Assays ›
          Gene Symbol:
          Dock9
          Gene Name:
          dedicator of cytokinesis 9
          Gene Aliases:
          Trg
          Chromosome Location:
          Chr.15: 107945664 - 108210985 on Build Rnor_6.0
          UniGene ID:
          Rn.10431
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS352405
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dock9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105759.2 NP_001099229.2 30, 31 3409
          XM_006252481.2 XP_006252543.1 30, 31 3520
          XM_006252488.2 XP_006252550.1 30, 31 3520
          XM_006252490.2 XP_006252552.1 30, 31 3520
          XM_006252493.2 XP_006252555.1 30, 31 3975
          XM_006252496.2 XP_006252558.1 30, 31 3544
          XM_006252499.2 XP_006252561.1 30, 31 3519
          XM_006252501.2 XP_006252563.1 30, 31 3519
          XM_006252502.2 XP_006252564.1 30, 31 3519
          XM_006252503.2 XP_006252565.1 30, 31 3568
          XM_006252505.2 XP_006252567.1 30, 31 3544
          XM_008770951.1 XP_008769173.1 30, 31 3520
          XM_008770952.1 XP_008769174.1 30, 31 3520
          XM_008770953.1 XP_008769175.1 30, 31 3520
          XM_008770954.1 XP_008769176.1 30, 31 3520
          XM_008770955.1 XP_008769177.1 30, 31 3545
          XM_008770956.1 XP_008769178.1 30, 31 3545
          XM_008770957.1 XP_008769179.1 30, 31 3520
          XM_008770958.1 XP_008769180.1 30, 31 3520
          XM_008770959.1 XP_008769181.1 30, 31 3520
          XM_008770960.1 XP_008769182.1 30, 31 3520
          XM_008770961.1 XP_008769183.1 30, 31 3520
          XM_008770962.1 XP_008769184.1 30, 31 3520
          XM_008770963.1 XP_008769185.1 30, 31 3520
          XM_008770964.1 XP_008769186.1 30, 31 3585
          XM_008770965.1 XP_008769187.1 30, 31 3520
          XM_008770966.1 XP_008769188.1 30, 31 3520
          XM_008770967.1 XP_008769189.1 30, 31 3519
          XM_008770968.1 XP_008769190.1 30, 31 3980
          XM_008770969.1 XP_008769191.1 30, 31 3981

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          Rho GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0