Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UCN SIRNA Assays ›
          Gene Symbol:
          Ucn
          Gene Name:
          urocortin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 26602144 - 26602974 on Build Rnor_6.0
          UniGene ID:
          Rn.10190
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS353875
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ucn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019150.1 NP_062023.1 2 231
          GenBank mRNA U33935.1 - 2 231

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          inflammatory response
          response to oxidative stress
          neuropeptide signaling pathway
          female pregnancy
          sensory perception of sound
          learning or memory
          feeding behavior
          associative learning
          aerobic respiration
          positive regulation of gene expression
          negative regulation of gene expression
          pancreatic juice secretion
          positive regulation of cell growth
          positive regulation of cAMP biosynthetic process
          negative regulation of histone deacetylation
          neuron projection development
          negative regulation of appetite
          response to estradiol
          positive regulation of interleukin-6 production
          positive regulation of collagen biosynthetic process
          positive regulation of peptidyl-serine phosphorylation
          activation of protein kinase A activity
          social behavior
          gastric emptying
          drinking behavior
          negative regulation of apoptotic process
          positive regulation of vascular permeability
          positive regulation of translation
          positive regulation of DNA replication
          negative regulation of blood pressure
          negative regulation of cell size
          positive regulation of vasodilation
          negative regulation of hormone secretion
          response to pain
          response to glucocorticoid
          positive regulation of corticotropin secretion
          regulation of synaptic transmission, glutamatergic
          positive regulation of cardiac muscle contraction
          negative regulation of gastric acid secretion
          negative regulation of necrotic cell death
          negative regulation of cell death
          positive regulation of calcium ion import
          negative regulation of neuron death
          negative regulation of feeding behavior
          positive regulation of behavioral fear response
          G-protein coupled receptor binding
          hormone activity
          protein binding
          histone deacetylase inhibitor activity
          corticotropin-releasing hormone receptor 1 binding
          corticotropin-releasing hormone receptor 2 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-fb85479f-282mh:80/100.66.130.62:80.
          git-commit: 1c69677c60948f80141da00d3c5091e873d99202
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline