Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013069.1 | NP_001013087.1 | 4 | 632 | ||
XM_006229850.2 | XP_006229912.1 | 5 | 755 | |||
XM_006229851.2 | XP_006229913.1 | 4 | 694 | |||
GenBank mRNA | BC091411.1 | - | 4 | 632 | 7325640 |