Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017196.3 | NP_058892.1 | 5 | 354 | ||
XM_006256061.2 | XP_006256123.1 | 4 | 728 | |||
XM_006256062.2 | XP_006256124.1 | 5 | 705 | |||
XM_006256063.2 | XP_006256125.1 | 5 | 668 | |||
XM_006256065.2 | XP_006256127.1 | 7 | 504 | |||
GenBank mRNA | AB000818.1 | - | 5 | 352 | ||
AF299328.1 | - | 2 | 94 | |||
BC081822.1 | - | 3 | 1201 | |||
D82069.1 | - | 7 | 509 | |||
FM053942.1 | - | 5 | 341 | |||
FQ231499.1 | - | 5 | 352 | |||
U10894.1 | - | 2 | 108 | |||
U17919.1 | - | 5 | 326 | |||
U33471.1 | - | 3 | 1378 |