Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199380.1 | NP_955412.1 | 8 | 862 | ||
GenBank mRNA | BC062402.1 | - | 8 | 862 | 5621303 | |
FQ213305.1 | - | 8 | 894 | |||
FQ215337.1 | - | 8 | 907 | |||
FQ215587.1 | - | 8 | 803 | |||
FQ234002.1 | - | 8 | 891 |