Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNNC2 SIRNA Assays ›
          Gene Symbol:
          Tnnc2
          Gene Name:
          troponin C type 2 (fast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 161243747 - 161244831 on Build Rnor_6.0
          UniGene ID:
          Rn.43529
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS355623
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tnnc2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037351.1 NP_001032428.1 4, 5 447
          GenBank mRNA DQ273679.1 - 4, 5 447
          FQ215069.1 - 511
          FQ216531.1 - 511
          FQ216869.1 - 511
          FQ216875.1 - 541
          FQ216940.1 - 503
          FQ216956.1 - 524
          FQ217047.1 - 511
          FQ217054.1 - 511
          FQ217106.1 - 540
          FQ217137.1 - 511
          FQ217169.1 - 511
          FQ217172.1 - 511
          FQ217195.1 - 511
          FQ217236.1 - 522
          FQ217299.1 - 511
          FQ217371.1 - 511
          FQ217378.1 - 511
          FQ217706.1 - 511
          FQ217788.1 - 511
          FQ217840.1 - 511
          FQ217842.1 - 511
          FQ217896.1 - 510
          FQ217974.1 - 540
          FQ217984.1 - 511
          FQ218010.1 - 511
          FQ218048.1 - 541
          FQ223543.1 - 511
          FQ223572.1 - 511
          FQ223583.1 - 511
          FQ223607.1 - 511
          FQ223623.1 - 511
          FQ223636.1 - 511
          FQ223643.1 - 511
          FQ223690.1 - 511
          FQ223692.1 - 511
          FQ223711.1 - 511
          FQ223713.1 - 511
          FQ223719.1 - 511
          FQ223773.1 - 523
          FQ223816.1 - 532
          FQ223825.1 - 511
          FQ223832.1 - 511
          FQ223835.1 - 541
          FQ223976.1 - 511
          FQ223986.1 - 511
          FQ224079.1 - 511
          FQ224114.1 - 540
          FQ224128.1 - 511
          FQ224223.1 - 511
          FQ224258.1 - 511
          FQ224340.1 - 511
          FQ224376.1 - 526
          FQ224378.1 - 511
          FQ224387.1 - 511
          FQ224450.1 - 511
          FQ224462.1 - 511
          FQ224599.1 - 511
          FQ224648.1 - 511
          FQ224672.1 - 511
          FQ224749.1 - 511
          FQ224755.1 - 540
          FQ224811.1 - 511
          FQ224822.1 - 511
          FQ224827.1 - 508

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          actin binding
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc