Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PINK1 SIRNA Assays ›
Gene Symbol:
Pink1
Gene Name:
PTEN induced putative kinase 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 156677146 - 156689258 on Build Rnor_6.0
UniGene ID:
Rn.219286
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS356200
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pink1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001106694.1
NP_001100164.1
4
780
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy
positive regulation of protein phosphorylation
protein phosphorylation
response to oxidative stress
mitochondrion organization
biological_process
regulation of hydrogen peroxide metabolic process
negative regulation of gene expression
negative regulation of macroautophagy
protein ubiquitination
peptidyl-serine phosphorylation
respiratory electron transport chain
regulation of protein ubiquitination
positive regulation of synaptic transmission, dopaminergic
positive regulation of peptidyl-serine phosphorylation
positive regulation of dopamine secretion
positive regulation of catecholamine secretion
cellular response to oxidative stress
positive regulation of protein dephosphorylation
intracellular signal transduction
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
regulation of protein complex assembly
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
positive regulation of translation
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of mitochondrial membrane potential
cellular response to hypoxia
positive regulation of mitochondrial fission
positive regulation of release of cytochrome c from mitochondria
negative regulation of mitochondrial fission
cellular response to toxic substance
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
negative regulation of autophagosome assembly
negative regulation of mitophagy
negative regulation of oxidative stress-induced cell death
regulation of protein targeting to mitochondrion
negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
negative regulation of hydrogen peroxide-induced neuron intrinsic apoptotic signaling pathway
positive regulation of cristae formation
regulation of reactive oxygen species metabolic process
negative regulation of reactive oxygen species metabolic process
magnesium ion binding
protease binding
molecular_function
protein serine/threonine kinase activity
protein binding
ATP binding
calcium-dependent protein kinase activity
kinase activity
ubiquitin protein ligase binding
protein kinase B binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-849d5b7584-dvwjr:80/100.66.130.62:80.
git-commit: 0a9b6045e9144c2c4a21b698be73aa131cdbda4b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0