Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSP90AA1 SIRNA Assays ›
          Gene Symbol:
          Hsp90aa1 LOC103692716
          Gene Name:
          heat shock protein 90, alpha (cytosolic), class A member 1
          heat shock protein HSP 90-alpha
          Gene Aliases:
          Hsp86, Hsp90, Hspca
          -
          Chromosome Location:
          Chr.6: 135107262 - 135112793 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 135044833 - 135050421 on Build Rnor_6.0
          UniGene ID:
          Rn.119867
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID RSS356414
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hsp90aa1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175761.2 NP_786937.1 4 616
          GenBank mRNA AJ428213.1 - 4 616
          BC072489.1 - 4 598 7105514
          BC085120.1 - 4 598 7114546
          FQ211921.1 - 4 616
          FQ213634.1 - 4 615
          FQ225235.1 - 4 772
          FQ225422.1 - 4 616
          FQ225565.1 - 4 616
          FQ225903.1 - 4 772
          FQ225992.1 - 4 616
          FQ226483.1 - 4 616
          FQ226612.1 - 4 617
          FQ227103.1 - 4 616
          FQ227479.1 - 4 616
          FQ227490.1 - 4 615
          FQ227525.1 - 4 617
          FQ228040.1 - 4 616
          FQ230359.1 - 4 616
          FQ230408.1 - 4 616
          FQ230565.1 - 4 616
          FQ230623.1 - 4 616
          FQ231096.1 - 4 616
          FQ231403.1 - 4 616
          FQ231559.1 - 4 620
          FQ231773.1 - 4 692
          FQ232301.1 - 4 772
          FQ232340.1 - 4 616
          FQ232738.1 - 4 616
          FQ232984.1 - 4 616
          FQ233436.1 - 4 616
          FQ233585.1 - 4 616
          FQ234090.1 - 4 616
          FQ234152.1 - 4 616
          FQ234628.1 - 4 616
          FQ235318.1 - 4 616
          Gene Symbol: LOC103692716
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008764969.1 XP_008763191.1 668

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          skeletal muscle contraction
          protein folding
          response to heat
          response to salt stress
          positive regulation of lamellipodium assembly
          cardiac muscle cell apoptotic process
          positive regulation of protein import into nucleus, translocation
          response to estrogen
          protein import into mitochondrial outer membrane
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell size
          regulation of catalytic activity
          chaperone-mediated protein complex assembly
          positive regulation of cardiac muscle contraction
          UTP binding
          CTP binding
          mRNA binding
          protein binding
          ATP binding
          GTP binding
          ATPase activity
          sulfonylurea receptor binding
          protein kinase binding
          protein phosphatase binding
          MHC class II protein complex binding
          nitric-oxide synthase regulator activity
          TPR domain binding
          dATP binding
          identical protein binding
          protein homodimerization activity
          ion channel binding
          poly(A) RNA binding
          Rho GDP-dissociation inhibitor binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0