Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126272.1 | NP_001119744.1 | 4 | 844 | ||
XM_006246383.2 | XP_006246445.1 | 9 | 1555 | |||
XM_006246384.2 | XP_006246446.1 | 9 | 1496 | |||
XM_006246385.2 | XP_006246447.1 | 9 | 1243 | |||
XM_006246386.2 | XP_006246448.1 | 9 | 1138 | |||
GenBank mRNA | BC166433.1 | - | 9 | 1129 | 7191618 |