Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037643.2 | NP_001032732.2 | 3 | 604 | ||
GenBank mRNA | BC107663.1 | - | 3 | 604 | 7936709 |