Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107202.2 | NP_001100672.1 | 8 | 1120 | ||
GenBank mRNA | EU448292.1 | - | 8 | 1120 | ||
EU448293.1 | - | 8 | 1271 |