Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751704.2 | XP_003751752.1 | 1 | 282 | ||
GenBank mRNA | BC098858.1 | - | 1 | 296 |