Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107684.1 | NP_001101154.1 | 18 | 3273 | ||
XM_006232511.2 | XP_006232573.2 | 23 | 5185 | |||
XM_006232512.2 | XP_006232574.1 | 21 | 4251 | |||
XM_006232513.2 | XP_006232575.1 | 21 | 4062 | |||
XM_008761092.1 | XP_008759314.1 | 27 | 4555 | |||
XM_008761093.1 | XP_008759315.1 | 26 | 4538 | |||
XM_008761094.1 | XP_008759316.1 | 26 | 4531 | |||
XM_008761095.1 | XP_008759317.1 | 27 | 4229 | |||
XM_008761096.1 | XP_008759318.1 | 25 | 4844 | |||
XM_008761097.1 | XP_008759319.1 | 24 | 4736 | |||
XM_008761098.1 | XP_008759320.1 | 24 | 6028 |